Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Spo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012046.2 | NP_036176.1 | 2 | 198 | ||
GenBank mRNA | AF169386.1 | - | 2 | 195 | ||
AF173848.1 | - | 2 | 191 |