Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013512.2 | NP_038540.2 | 3 | 897 | ||
GenBank mRNA | AK140485.1 | - | 2 | 581 | ||
AK161182.1 | - | 3 | 657 | |||
BC007166.1 | - | 3 | 621 | 3490061 | ||
BC011099.1 | - | 2 | 216 | |||
BC013557.1 | - | 3 | 621 | 3490326 | ||
BC089158.1 | - | 2 | 488 | |||
D28818.1 | - | 2 | 171 |