Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Itgb1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013712.2 | NP_038740.1 | 11 | 1234 | ||
GenBank mRNA | AF140691.1 | - | 11 | 1178 | ||
AK003906.1 | - | 11 | 1235 | |||
BC030035.1 | - | 11 | 1209 | 1346742 |