Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013725.4 | NP_038753.1 | 3 | 242 | ||
GenBank mRNA | AK005147.1 | - | 3 | 187 | ||
AK011207.1 | - | 745 | ||||
AK150738.1 | - | 3 | 217 | |||
BC012641.1 | - | 3 | 256 | 4019309 | ||
U93864.1 | - | 3 | 172 |