Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RGS14 SIRNA Assays ›
Gene Symbol:
Rgs14
Gene Name:
regulator of G-protein signaling 14
Gene Aliases:
RPIP1
Chromosome Location:
Chr.13: 55369712 - 55384691 on Build GRCm38
UniGene ID:
Mm.1426
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
72418
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rgs14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_016758.3
NP_058038.2
4
429
GenBank mRNA
AK154944.1
-
4
354
BC030321.1
-
4
401
5374531
U85055.1
-
4
269
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nucleocytoplasmic transport
response to oxidative stress
cell cycle
spindle organization
chromosome segregation
mitotic nuclear division
signal transduction
G-protein coupled receptor signaling pathway
multicellular organism development
learning
long-term memory
regulation of G-protein coupled receptor protein signaling pathway
visual learning
negative regulation of signal transduction
zygote asymmetric cell division
negative regulation of synaptic plasticity
intracellular signal transduction
termination of G-protein coupled receptor signaling pathway
negative regulation of MAP kinase activity
positive regulation of GTPase activity
regulation of DNA-templated transcription in response to stress
platelet-derived growth factor receptor signaling pathway
positive regulation of neurogenesis
cell division
long-term synaptic potentiation
negative regulation of ERK1 and ERK2 cascade
G-protein alpha-subunit binding
receptor signaling protein activity
GDP-dissociation inhibitor activity
GTPase activator activity
protein binding
microtubule binding
protein kinase binding
receptor signaling complex scaffold activity
GTPase regulator activity
GTPase activating protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0