Search
Search

Pdlim1| Gene Symbol: Pdlim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016861.4 | NP_058557.2 | 2 | 297 | ||
| GenBank mRNA | AF053367.2 | - | 2 | 213 | ||
| AK076285.1 | - | 196 | ||||
| AK088089.1 | - | 2 | 196 | |||
| AK146225.1 | - | 2 | 207 | |||
| AK146399.1 | - | 2 | 209 | |||
| AK158173.1 | - | 2 | 196 | |||
| AK158992.1 | - | 2 | 297 | |||
| BC004809.1 | - | 2 | 188 | 3588132 | ||