Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: PARP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006437.3 | NP_006428.2 | 2 | 207 | ||
XM_011534932.2 | XP_011533234.1 | 2 | 424 | |||
GenBank mRNA | AF057160.1 | - | 2 | 153 | ||
AF158255.1 | - | 2 | 207 | |||
AK304399.1 | - | 2 | 175 | |||
BC017815.1 | - | 2 | 146 | |||
D79999.2 | - | 1 | 106 | |||
DB238428.1 | - | 2 | 175 |