Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNRNPC SIRNA Assays ›
          Gene Symbol:
          Hnrnpc
          Gene Name:
          heterogeneous nuclear ribonucleoprotein C
          Gene Aliases:
          AL022939, D14Wsu171e, Hnrpc, Hnrpc1, Hnrpc2, hnRNPC1, hnRNPC2, hnrnp-C
          Chromosome Location:
          Chr.14: 52073380 - 52104028 on Build GRCm38
          UniGene ID:
          Mm.427321
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 72792

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hnrnpc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001170981.1 NP_001164452.1 3 420
          NM_001170982.1 NP_001164453.1 2 394
          NM_001170983.1 NP_001164454.1 3 420
          NM_001170984.1 NP_001164455.1 3 420
          NM_016884.3 NP_058580.1 3 420
          GenBank mRNA AF095257.1 - 3 374
          AK010845.1 - 3 399
          AK011336.1 - 3 392
          AK012633.2 - 3 382
          AK019958.1 - 3 1595
          AK088245.1 - 3 397
          AK088678.1 - 3 392
          AK089061.1 - 3 400
          AK150399.1 - 3 396
          AK151446.1 - 3 397
          AK153053.1 - 3 397
          AK166590.1 - 3 398
          AK168802.1 - 3 394
          AK169287.1 - 3 392
          AK184811.1 - 3 327
          AK212671.1 - 1 246
          AK214960.1 - 2 282
          BC004706.1 - 3 382 3499283
          BC095922.1 - 2 335 4988965
          CK793036.1 - 374
          DV652725.1 - 3 444

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001077442, NM_001013631, NM_001146181, NM_001136561, NM_031314, and others.
          This siRNA also targets the following transcripts in Rat: NM_001277601, NM_001277602, NM_001271241, NM_001277600, XM_006251861, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          osteoblast differentiation
          mRNA processing
          RNA splicing
          ATP-dependent chromatin remodeling
          3'-UTR-mediated mRNA stabilization
          nucleotide binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          nucleic acid binding
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          poly(U) RNA binding
          nucleosomal DNA binding
          identical protein binding
          poly(A) RNA binding
          N6-methyladenosine-containing RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0