Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rragc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017475.2 | NP_059503.2 | 1, 2 | 343 | ||
GenBank mRNA | AB017616.1 | - | 1, 2 | 266 | ||
AK150792.1 | - | 1, 2 | 297 | |||
AK159355.1 | - | 1, 2 | 276 | |||
AK159434.1 | - | 1, 2 | 298 | |||
AK166658.1 | - | 1, 2 | 289 | |||
BC005417.1 | - | 1, 2 | 291 | 3495022 | ||
BC071245.1 | - | 1, 2 | 302 | |||
CJ172014.1 | - | 1, 2 | 344 |