Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vpreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016982.2 | NP_058678.1 | 2 | 742 | ||
GenBank mRNA | AK010843.1 | - | 2 | 736 | ||
AK010879.1 | - | 2 | 736 | |||
X05556.1 | - | 2 | 737 |