Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019626.3 | NP_062600.2 | 3 | 1471 | ||
GenBank mRNA | AK158818.1 | - | 3 | 1472 | ||
AK158885.1 | - | 3 | 1489 | |||
AK161846.1 | - | 3 | 1070 | |||
BC051956.1 | - | 3 | 1087 | |||
BC055730.1 | - | 3 | 1175 | 6404555 | ||
BC132120.1 | - | 4 | 1154 | 40130397 | ||
BC132122.1 | - | 4 | 1221 | 40130399 | ||
X61448.1 | - | 2 | 989 |