Search
Search

Nme3| Gene Symbol: Nme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019730.2 | NP_062704.2 | 4, 5 | 406 | ||
| GenBank mRNA | AF153449.1 | - | 4, 5 | 418 | ||
| AF288689.1 | - | 4, 5 | 400 | |||
| AK007399.1 | - | 4, 5 | 406 | |||
| BC028503.1 | - | 4, 5 | 397 | 3326648 | ||