Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLCO1B2 SIRNA Assays ›
Gene Symbol:
Slco1b2
Gene Name:
solute carrier organic anion transporter family, member 1b2
Gene Aliases:
7330442B20Rik, OATP-C, OATP2, Oatp1b2, Oatp4, Slc21a10, Slc21a6, lst-1, mlst-1
Chromosome Location:
Chr.6: 141629452 - 141686646 on Build GRCm38
UniGene ID:
Mm.272223
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
74958
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Slco1b2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_020495.1
NP_065241.1
3, 4
282
GenBank mRNA
AB031959.1
-
3, 4
282
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
drug transmembrane transport
oligopeptide transport
organic anion transport
bile acid and bile salt transport
drug transport
anion transmembrane transport
organic acid transmembrane transport
transporter activity
organic anion transmembrane transporter activity
bile acid transmembrane transporter activity
oligopeptide transporter activity
drug transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0