Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KCNK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317090.1 | NP_001304019.1 | 5 | 797 | ||
NM_033310.2 | NP_201567.1 | 5 | 934 | |||
GenBank mRNA | AF248242.1 | - | 4 | 971 | ||
AF259500.1 | - | 5 | 852 | |||
AK296021.1 | - | 3 | 686 | |||
BC110327.1 | - | 5 | 797 | |||
EU978935.1 | - | 4 | 630 |
Gene Symbol: KCNK4-TEX40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133662.1 | 5 | 752 |