Search
Search

SCN3B| Gene Symbol: SCN3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040151.1 | NP_001035241.1 | 3 | 616 | ||
| NM_018400.3 | NP_060870.1 | 2 | 1015 | |||
| XM_011542897.1 | XP_011541199.1 | 3 | 616 | |||
| GenBank mRNA | AB032984.1 | - | 3 | 256 | ||
| AJ243396.2 | - | 2 | 584 | |||
| AK314513.1 | - | 3 | 603 | |||
| AL136589.1 | - | 2 | 1012 | |||
| BC117282.1 | - | 2 | 302 | 40125833 | ||
| BC126265.1 | - | 2 | 302 | 8991981 | ||