Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RSAD2 SIRNA Assays ›
          Gene Symbol:
          Rsad2
          Gene Name:
          radical S-adenosyl methionine domain containing 2
          Gene Aliases:
          2510004L01Rik, Vig1, cig5
          Chromosome Location:
          Chr.12: 26442743 - 26456478 on Build GRCm38
          UniGene ID:
          Mm.24045
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 75666

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rsad2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021384.4 NP_067359.2 6 1630
          GenBank mRNA AK010906.1 - 6 1609
          AK036373.1 - 6 1610
          AK149900.1 - 6 1611
          AK150069.1 - 6 1612
          AK150347.1 - 6 1487
          AK150406.1 - 6 1610
          AK150425.1 - 6 1612
          AK150525.1 - 6 1611
          AK150536.1 - 6 1613
          AK150616.1 - 6 1610
          AK150684.1 - 6 1612
          AK150731.1 - 6 1612
          AK150767.1 - 6 1612
          AK150783.1 - 6 1612
          AK150788.1 - 6 1612
          AK150812.1 - 6 1612
          AK150835.1 - 6 1611
          AK151140.1 - 6 1612
          AK151185.1 - 6 1611
          AK151191.1 - 6 1612
          AK151206.1 - 6 1612
          AK151286.1 - 6 1610
          AK151308.1 - 6 1612
          AK151331.1 - 7 2047
          AK151339.1 - 6 1610
          AK151389.1 - 6 1612
          AK151410.1 - 6 1611
          AK151412.1 - 6 1610
          AK151414.1 - 6 1611
          AK151476.1 - 6 1612
          AK151502.1 - 6 1612
          AK151538.1 - 6 1612
          AK151767.1 - 6 1612
          AK151770.1 - 6 1610
          AK151825.1 - 6 1610
          AK151833.1 - 6 1610
          AK151846.1 - 6 1610
          AK151906.1 - 6 1612
          AK151909.1 - 6 1610
          AK151940.1 - 6 1610
          AK151954.1 - 6 1612
          AK151971.1 - 6 1612
          AK152050.1 - 6 1610
          AK152065.1 - 6 1612
          AK152075.1 - 6 1612
          AK152235.1 - 6 1610
          AK152239.1 - 6 1612
          AK152241.1 - 6 1612
          AK152310.1 - 6 1610
          AK152354.1 - 6 1610
          AK152493.1 - 6 1612
          AK152600.1 - 6 1610
          AK152601.1 - 6 1612
          AK152631.1 - 6 1610
          AK152644.1 - 6 1612
          AK152664.1 - 6 1612
          AK152701.1 - 6 1610
          AK152796.1 - 6 1610
          AK152861.1 - 6 1610
          AK152880.1 - 6 1610
          AK152895.1 - 6 1612
          AK152934.1 - 6 1612
          AK152962.1 - 6 1610
          AK153003.1 - 6 1612
          AK153024.1 - 6 1612
          AK153062.1 - 6 1614
          AK153077.1 - 6 1610
          AK153078.1 - 6 1610
          AK153232.1 - 6 1612
          AK153234.1 - 6 1612
          AK153306.1 - 6 1610
          AK153318.1 - 6 1612
          AK153324.1 - 6 1612
          AK153350.1 - 7 2047
          AK153435.1 - 6 1488
          AK153453.1 - 6 1576
          AK153556.1 - 6 1612
          AK159147.1 - 6 1610
          BC057868.1 - 6 1603 5327504

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          immune system process
          response to virus
          regulation of ossification
          positive regulation of toll-like receptor 7 signaling pathway
          positive regulation of toll-like receptor 9 signaling pathway
          CD4-positive, alpha-beta T cell activation
          CD4-positive, alpha-beta T cell differentiation
          negative regulation of viral genome replication
          innate immune response
          negative regulation of protein secretion
          defense response to virus
          positive regulation of T-helper 2 cell cytokine production
          catalytic activity
          protein binding
          protein self-association
          metal ion binding
          iron-sulfur cluster binding
          4 iron, 4 sulfur cluster binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0