Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pdcd1lg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021396.2 | NP_067371.1 | 6 | 1426 | ||
GenBank mRNA | AF142780.2 | - | 6 | 1416 | ||
AK089369.1 | - | 6 | 1429 | |||
AK141111.1 | - | 5 | 1516 | |||
AK155002.1 | - | 6 | 1366 |