Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021535.4 | NP_067510.3 | 2 | 184 | ||
GenBank mRNA | AB044414.1 | - | 2 | 113 | ||
AK003493.1 | - | 2 | 171 | |||
AK011140.1 | - | 2 | 183 | |||
AK019174.1 | - | 2 | 143 | |||
AK145982.1 | - | 2 | 183 | |||
AK159635.1 | - | 2 | 171 | |||
AK168813.1 | - | 2 | 178 | |||
BC020034.1 | - | 2 | 126 | |||
BC057446.1 | - | 2 | 145 | 3155760 | ||
BY036103.1 | - | 2 | 186 |