Search
Search

Fignl1| Gene Symbol: Fignl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163359.1 | NP_001156831.1 | 3 | 393 | ||
| NM_001163360.1 | NP_001156832.1 | 2 | 263 | |||
| NM_021891.3 | NP_068691.2 | 3 | 383 | |||
| GenBank mRNA | AF263914.1 | - | 2 | 297 | ||
| AK051324.1 | - | 3 | 491 | |||
| AK051874.1 | - | 3 | 353 | |||
| AK082092.1 | - | 347 | ||||
| AK086769.1 | - | 3 | 381 | |||
| AK088574.1 | - | 3 | 366 | |||
| AK143850.1 | - | 3 | 362 | |||
| AK148994.1 | - | 1 | 416 | |||
| BC051942.1 | - | 3 | 336 | 6401467 | ||
| BC052415.1 | - | 3 | 336 | 6401467 | ||
| BY231020.1 | - | 2 | 229 | |||
| CN693395.1 | - | 3 | 506 | |||