Search
Search

Kif23| Gene Symbol: Kif23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024245.4 | NP_077207.3 | 3, 4 | 392 | ||
| GenBank mRNA | AK013939.1 | - | 3, 4 | 330 | ||
| AK030771.1 | - | 3, 4 | 369 | |||
| BC003363.1 | - | 3, 4 | 365 | |||
| BC047273.1 | - | 3, 4 | 350 | 6440797 | ||