Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEF2C SIRNA Assays ›
          Gene Symbol:
          Mef2c
          Gene Name:
          myocyte enhancer factor 2C
          Gene Aliases:
          5430401D19Rik, 9930028G15Rik, AV011172, Mef2
          Chromosome Location:
          Chr.13: 83504034 - 83667079 on Build GRCm38
          UniGene ID:
          Mm.24001
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 77710

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mef2c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001170537.1 NP_001164008.1 4 657
          NM_025282.3 NP_079558.1 4 657
          GenBank mRNA AK009139.1 - 4 497
          AK047994.1 - 3 380
          AK077603.1 - 3 446
          AK132573.1 - 4 486
          BC026841.1 - 3 389 4481227
          BC037731.1 - 3 385 4500786
          BC057650.1 - 3 374 4500786
          L13171.1 - 2 218

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          MADS box transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          blood vessel development
          osteoblast differentiation
          neuron migration
          B cell homeostasis
          heart looping
          endochondral ossification
          blood vessel remodeling
          chondrocyte differentiation
          germinal center formation
          regulation of germinal center formation
          response to ischemia
          primary heart field specification
          secondary heart field specification
          outflow tract morphogenesis
          sinoatrial valve morphogenesis
          cardiac ventricle formation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          apoptotic process
          humoral immune response
          multicellular organism development
          nervous system development
          heart development
          skeletal muscle tissue development
          muscle cell fate determination
          learning or memory
          positive regulation of cardiac muscle hypertrophy
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of alkaline phosphatase activity
          neural crest cell differentiation
          cardiac muscle hypertrophy in response to stress
          cell differentiation
          neuron differentiation
          platelet formation
          monocyte differentiation
          negative regulation of ossification
          melanocyte differentiation
          positive regulation of bone mineralization
          positive regulation of B cell proliferation
          cellular response to drug
          skeletal muscle cell differentiation
          cellular response to trichostatin A
          B cell proliferation
          positive regulation of MAP kinase activity
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          cell fate commitment
          regulation of megakaryocyte differentiation
          positive regulation of myoblast differentiation
          positive regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of neurotransmitter secretion
          regulation of synaptic plasticity
          positive regulation of skeletal muscle tissue development
          neuron development
          cell morphogenesis involved in neuron differentiation
          embryonic viscerocranium morphogenesis
          embryonic skeletal system morphogenesis
          negative regulation of epithelial cell proliferation
          B cell receptor signaling pathway
          smooth muscle cell differentiation
          regulation of synapse assembly
          regulation of synaptic transmission, glutamatergic
          cardiac muscle cell differentiation
          ventricular cardiac muscle cell differentiation
          palate development
          regulation of synaptic activity
          positive regulation of cardiac muscle cell proliferation
          excitatory postsynaptic potential
          transdifferentiation
          regulation of sarcomere organization
          cartilage morphogenesis
          regulation of dendritic spine development
          renal tubule morphogenesis
          cellular response to lipopolysaccharide
          cellular response to calcium ion
          cellular response to retinoic acid
          cellular response to growth factor stimulus
          cellular response to parathyroid hormone stimulus
          cellular response to fluid shear stress
          cellular response to transforming growth factor beta stimulus
          positive regulation of cell proliferation in bone marrow
          glomerulus morphogenesis
          nephron tubule epithelial cell differentiation
          positive regulation of protein homodimerization activity
          positive regulation of macrophage apoptotic process
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          positive regulation of cardiac muscle cell differentiation
          positive regulation of behavioral fear response
          epithelial cell proliferation involved in renal tubule morphogenesis
          positive regulation of skeletal muscle cell differentiation
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter sequence-specific
          core promoter proximal region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          DNA binding
          AT DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          activating transcription factor binding
          miRNA binding
          histone deacetylase binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity
          protein dimerization activity
          HMG box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0