Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rmnd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024288.2 | NP_077250.2 | 9 | 3225 | ||
GenBank mRNA | AK083326.1 | - | 3 | 1863 | ||
AK087966.1 | - | 3 | 1866 | |||
AK088356.1 | - | 3 | 1866 | |||
AK137858.1 | - | 1 | 1166 | |||
AK162772.1 | - | 3 | 1877 | |||
BC034085.1 | - | 6 | 1086 | |||
BC050876.1 | - | 9 | 3076 | 6512227 |