Search
Search

Haus2| Gene Symbol: Haus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290807.1 | NP_001277736.1 | 6 | 749 | ||
| NM_001290808.1 | NP_001277737.1 | 5 | 779 | |||
| NM_025475.3 | NP_079751.1 | 6 | 849 | |||
| GenBank mRNA | AK012086.1 | - | 6 | 848 | ||
| AK012779.1 | - | 5 | 779 | |||
| AK012960.1 | - | 6 | 796 | |||
| AK132300.1 | - | 6 | 805 | |||
| BC086677.1 | - | 6 | 787 | 30660268 | ||