Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prl2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025532.3 | NP_079808.1 | 5 | 726 | ||
GenBank mRNA | AF525155.1 | - | 5 | 677 | ||
AK005411.1 | - | 5 | 727 | |||
AY077703.1 | - | 1 | 154 | |||
BC100329.1 | - | 5 | 716 | 30787440 | ||
BC111112.1 | - | 5 | 722 | 40050224 | ||
BC138199.1 | - | 5 | 719 | 8861218 |