Search
Search

Trappc5| Gene Symbol: Trappc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025701.4 | NP_079977.3 | 2 | 1884 | ||
| GenBank mRNA | AK076212.1 | - | 2 | 1885 | ||
| AK078451.1 | - | 2 | 1850 | |||
| AK171833.1 | - | 1 | 4264 | |||
| BC028946.1 | - | 1 | 2344 | |||
| BC049179.1 | - | 2 | 1842 | 6313540 | ||