Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102407.1 | NP_001095877.1 | 3 | 198 | ||
NM_025875.2 | NP_080151.2 | 3 | 195 | |||
GenBank mRNA | AK009953.1 | - | 3 | 190 | ||
AK010284.1 | - | 3 | 183 | |||
AK082925.1 | - | 3 | 187 | |||
BC020086.1 | - | 3 | 178 | 3671644 | ||
BC058376.1 | - | 3 | 167 | 5709938 | ||
BY128307.1 | - | 3 | 192 | |||
CJ072219.1 | - | 3 | 199 | |||
CJ075560.1 | - | 3 | 199 | |||
KF051021.1 | - | 3 | 164 |