Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WWP2 SIRNA Assays ›
          Gene Symbol:
          Wwp2
          Gene Name:
          WW domain containing E3 ubiquitin protein ligase 2
          Gene Aliases:
          1300010O06Rik, AA690238, AIP2, AW554328
          Chromosome Location:
          Chr.8: 107436368 - 107558597 on Build GRCm38
          UniGene ID:
          Mm.390058
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 79141

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Wwp2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_025830.3 NP_080106.1 4 342
          GenBank mRNA AK004962.2 - 4 358
          AK088936.1 - 4 321
          AK141281.1 - 4 352
          AK146306.1 - 4 342
          AK151548.1 - 4 336
          BC039921.1 - 4 334 4954925
          BC048184.1 - 4 342 6827381
          JN712751.1 - 4 375

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106184, XM_008772560, XM_008772561, BC168152, JN712752.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          negative regulation of gene expression
          protein ubiquitination
          negative regulation of transporter activity
          regulation of ion transmembrane transport
          regulation of membrane potential
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of protein transport
          protein autoubiquitination
          protein K63-linked ubiquitination
          regulation of potassium ion transmembrane transporter activity
          RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          ubiquitin-protein transferase activity
          protein binding
          transcription factor binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          ubiquitin protein ligase activity
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0