Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102407.1 | NP_001095877.1 | 4 | 256 | ||
NM_025875.2 | NP_080151.2 | 4 | 253 | |||
GenBank mRNA | AK009953.1 | - | 4 | 248 | ||
AK010284.1 | - | 4 | 241 | |||
AK082925.1 | - | 4 | 245 | |||
BC020086.1 | - | 4 | 236 | 3671644 | ||
BC058376.1 | - | 4 | 225 | 5709938 | ||
BY128307.1 | - | 4 | 250 | |||
KF051021.1 | - | 4 | 222 |
Gene Symbol: B020018G12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139805.1 | - | 1 | 430 | ||
BC132651.1 | - | 1 | 419 | 40130928 | ||
BC132653.1 | - | 1 | 419 | 40130930 |