Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pqlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164420.1 | NP_001157892.1 | 5 | 1929 | ||
NM_001164421.1 | NP_001157893.1 | 5 | 1861 | |||
NM_001164422.1 | NP_001157894.1 | 5 | 1856 | |||
NM_025861.3 | NP_080137.2 | 6 | 1915 | |||
GenBank mRNA | AK009256.1 | - | 5 | 1851 | ||
AK018759.1 | - | 5 | 2293 | |||
AK153023.1 | - | 5 | 1883 | |||
AK153176.1 | - | 5 | 1882 | |||
BB728808.1 | - | 1 | 352 | |||
BC008231.1 | - | 5 | 1798 | 2647794 |