Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prl7c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026206.2 | NP_080482.1 | 4 | 330 | ||
GenBank mRNA | AF525157.1 | - | 4 | 315 | ||
AK005486.1 | - | 4 | 331 | |||
AK014436.1 | - | 4 | 314 | |||
BC099442.1 | - | 4 | 290 | 30716548 |