Search
Search

Necap1| Gene Symbol: Necap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026267.2 | NP_080543.2 | 8 | 2054 | ||
| GenBank mRNA | AK004805.1 | - | 8 | 2055 | ||
| AK005263.1 | - | 8 | 2045 | |||
| AK081598.1 | - | 8 | 2055 | |||
| AK159237.1 | - | 1 | 1164 | |||
| AK168500.1 | - | 8 | 2055 | |||
| AK220236.1 | - | 8 | 2038 | |||