Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VRK2 SIRNA Assays ›
Gene Symbol:
VRK2
Gene Name:
vaccinia related kinase 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 57907651 - 58163991 on Build GRCh38
UniGene ID:
Hs.715298
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
805
Purification
In-Vivo Ready
Standard
HPLC
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
6,301.00
Your Price
Online offer:
6301.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VRK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130480.2
NP_001123952.1
2
269
NM_001130481.2
NP_001123953.1
2
111
NM_001130482.2
NP_001123954.1
2
215
NM_001130483.2
NP_001123955.1
2
269
NM_001288836.1
NP_001275765.1
2
269
NM_001288837.1
NP_001275766.1
5
829
NM_001288838.1
NP_001275767.1
4
711
NM_001288839.1
NP_001275768.1
2
316
NM_006296.6
NP_006287.2
2
316
XM_005264540.4
XP_005264597.1
2
329
XM_006712090.3
XP_006712153.1
2
124
XM_006712091.3
XP_006712154.1
2
364
XM_006712092.3
XP_006712155.1
2
364
XM_006712093.3
XP_006712156.1
2
364
XM_011533092.2
XP_011531394.1
2
113
XM_017004858.1
XP_016860347.1
2
269
XM_017004859.1
XP_016860348.1
2
364
XM_017004860.1
XP_016860349.1
2
364
XM_017004861.1
XP_016860350.1
2
178
XM_017004862.1
XP_016860351.1
2
364
XM_017004863.1
XP_016860352.1
2
364
XM_017004864.1
XP_016860353.1
2
364
XM_017004865.1
XP_016860354.1
2
178
XM_017004866.1
XP_016860355.1
2
178
GenBank mRNA
AB000450.1
-
2
214
AJ512204.2
-
2
214
AK058199.1
-
2
171
AK223540.1
-
2
171
AK292727.1
-
4
697
AK295193.1
-
2
149
AK296611.1
-
2
254
AY228367.1
-
2
111
AY228368.1
-
2
246
AY228369.1
-
2
144
AY228370.1
-
2
199
AY228371.1
-
2
246
AY228372.1
-
2
199
BC021663.1
-
2
142
BC027854.1
-
2
267
5221315
BI906749.1
-
2
276
DB227029.1
-
2
269
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
regulation of cell shape
viral process
peptidyl-serine phosphorylation
cellular response to oxidative stress
regulation of MAPK cascade
protein autophosphorylation
regulation of interleukin-1-mediated signaling pathway
protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0