Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cdca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026410.3 | NP_080686.1 | 5 | 356 | ||
GenBank mRNA | AK010540.1 | - | 5 | 323 | ||
AK011701.1 | - | 5 | 356 | |||
AK016305.1 | - | 5 | 470 | |||
BC029626.1 | - | 4 | 250 | |||
BC052904.1 | - | 5 | 354 | 30065848 |