Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Murc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026509.3 | NP_080785.2 | 2 | 638 | ||
GenBank mRNA | AK009691.1 | - | 2 | 595 | ||
BC089601.1 | - | 2 | 638 | 6788090 | ||
EU487254.1 | - | 2 | 530 |