Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cgnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018850.1 | - | 3 | 615 | ||
AK019857.1 | - | 3 | 602 | |||
AK173252.1 | - | 17 | 2839 | |||
BC031499.1 | - | 3 | 451 | |||
BC039211.1 | - | 11 | 1557 | |||
BC139006.1 | - | 18 | 3983 | 8862028 | ||
BC158061.1 | - | 18 | 3986 | 9007361 |