Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYO7A SIRNA Assays ›
          Gene Symbol:
          MYO7A
          Gene Name:
          myosin VIIA
          Gene Aliases:
          DFNA11, DFNB2, MYOVIIA, MYU7A, NSRD2, USH1B
          Chromosome Location:
          Chr.11: 77128192 - 77215241 on Build GRCh38
          UniGene ID:
          Hs.370421
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 8175

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYO7A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000260.3 NP_000251.3 8 1029
          NM_001127179.2 NP_001120651.2 8 1029
          NM_001127180.1 NP_001120652.1 8 1023
          XM_011545044.2 XP_011543346.1 8 1101
          XM_011545046.2 XP_011543348.2 7 862
          XM_011545050.2 XP_011543352.1 5 716
          XM_017017778.1 XP_016873267.1 7 862
          XM_017017779.1 XP_016873268.1 7 862
          XM_017017780.1 XP_016873269.1 7 862
          XM_017017781.1 XP_016873270.1 7 862
          XM_017017782.1 XP_016873271.1 7 862
          XM_017017783.1 XP_016873272.1 7 862
          XM_017017784.1 XP_016873273.1 7 862
          XM_017017785.1 XP_016873274.1 7 862
          XM_017017786.1 XP_016873275.1 7 862
          XM_017017787.1 XP_016873276.1 7 862
          XM_017017788.1 XP_016873277.1 7 862
          GenBank mRNA BC144519.1 - 9 1071
          U34227.1 - 8 1024
          U39226.1 - 8 1029
          U55208.1 - 8 1024
          U55209.1 - 8 1024

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          phagolysosome assembly
          intracellular protein transport
          lysosome organization
          visual perception
          sensory perception of sound
          actin filament-based movement
          eye photoreceptor cell development
          post-embryonic organ morphogenesis
          sensory perception of light stimulus
          equilibrioception
          pigment granule transport
          auditory receptor cell stereocilium organization
          microfilament motor activity
          protein binding
          calmodulin binding
          ATP binding
          protein domain specific binding
          spectrin binding
          actin-dependent ATPase activity
          protein homodimerization activity
          ADP binding
          protein N-terminus binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0