Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 5430416O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017317.1 | - | 2 | 925 | ||
AK030334.1 | - | 2 | 943 | |||
BC064043.1 | - | 2 | 910 | |||
BC147344.1 | - | 1 | 474 | 9056418 | ||
BC147345.1 | - | 1 | 474 | 9056411 |