Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Syce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168244.1 | NP_001161716.1 | 2 | 656 | ||
NM_001168246.1 | NP_001161718.1 | 3 | 355 | |||
NM_027954.3 | NP_082230.2 | 3 | 337 | |||
GenBank mRNA | AK005954.1 | - | 3 | 338 | ||
AK167332.1 | - | 2 | 655 | |||
BC094628.1 | - | 3 | 232 | 3374002 | ||
BC132534.1 | - | 3 | 287 | 40130811 | ||
BY246600.1 | - | 3 | 270 |