Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Jmjd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028101.4 | NP_082377.2 | 9 | 1462 | ||
GenBank mRNA | AK011308.1 | - | 9 | 1459 | ||
AK148079.1 | - | 9 | 1440 | |||
AK155810.1 | - | 9 | 1464 | |||
AK172073.1 | - | 9 | 1459 | |||
BC010800.1 | - | 9 | 1424 | 4235072 |