Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rab38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028238.7 | NP_082514.4 | 2 | 531 | ||
GenBank mRNA | AB232634.1 | - | 2 | 402 | ||
AK009296.1 | - | 2 | 492 | |||
AK044783.1 | - | 2 | 494 | |||
AK054253.1 | - | 2 | 532 | |||
AK165682.1 | - | 2 | 534 | |||
AK196794.1 | - | 1 | 162 | |||
AK197116.1 | - | 1 | 162 | |||
AY062237.1 | - | 2 | 491 | |||
BC089578.1 | - | 2 | 491 | 6762755 |