Search
Search

NAGLU| Gene Symbol: NAGLU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000263.3 | NP_000254.2 | 5 | 1125 | ||
| XM_017024686.1 | XP_016880175.1 | 4 | 658 | |||
| GenBank mRNA | BC053991.1 | - | 5 | 808 | 6213852 | |
| L78464.1 | - | 5 | 791 | |||
| U40846.1 | - | 5 | 886 | |||
| U43573.1 | - | 5 | 785 | |||