Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tgm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028799.2 | NP_083075.1 | 3 | 325 | ||
GenBank mRNA | AK009196.2 | - | 3 | 325 | ||
AK037074.1 | - | 3 | 379 | |||
AK079431.1 | - | 3 | 389 | |||
AK132482.1 | - | 3 | 408 | |||
BC116266.1 | - | 3 | 308 | 40094000 | ||
BC116267.1 | - | 3 | 308 | 40094002 |