Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Spag17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028892.4 | NP_083168.3 | 7 | 959 | ||
GenBank mRNA | AK016477.1 | - | 7 | 945 | ||
AK029691.1 | - | 7 | 960 | |||
AK029947.1 | - | 7 | 955 | |||
AY555275.1 | - | 7 | 945 | |||
AY792594.1 | - | 7 | 945 |