Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POMK SIRNA Assays ›
          Gene Symbol:
          Pomk
          Gene Name:
          protein-O-mannose kinase
          Gene Aliases:
          4930444A02Rik, Sgk196
          Chromosome Location:
          Chr.8: 25980604 - 25994121 on Build GRCm38
          UniGene ID:
          Mm.17631
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 83581

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pomk
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_029037.4 NP_083313.1 5 712
          GenBank mRNA AK015374.1 - 5 705
          AK032677.1 - 5 713
          AK036348.1 - 5 719
          AK043620.1 - 5 713
          AK138952.1 - 5 721
          AK160873.1 - 5 718
          AK170994.1 - 5 713
          BC027296.1 - 4 692 4013355

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          protein phosphorylation
          protein O-linked glycosylation
          brain development
          learning or memory
          phosphorylation
          sensory perception of pain
          carbohydrate phosphorylation
          neuromuscular process
          nucleotide binding
          protein kinase activity
          ATP binding
          phosphofructokinase activity
          phosphorylase kinase regulator activity
          cobinamide kinase activity
          phytol kinase activity
          kinase activity
          phosphatidylinositol phosphate kinase activity
          cyclin-dependent protein serine/threonine kinase regulator activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          phosphotransferase activity, alcohol group as acceptor
          phenol kinase activity
          carbohydrate kinase activity
          cyclin-dependent protein kinase activating kinase regulator activity
          inositol tetrakisphosphate 2-kinase activity
          heptose 7-phosphate kinase activity
          aminoglycoside phosphotransferase activity
          phosphatidylinositol 3-kinase activity
          eukaryotic elongation factor-2 kinase regulator activity
          eukaryotic elongation factor-2 kinase activator activity
          LPPG:FO 2-phospho-L-lactate transferase activity
          cytidine kinase activity
          glycerate 2-kinase activity
          (S)-lactate 2-kinase activity
          phosphoserine:homoserine phosphotransferase activity
          L-seryl-tRNA(Sec) kinase activity
          phosphocholine transferase activity
          polynucleotide 5'-hydroxyl-kinase activity
          ATP-dependent polynucleotide kinase activity
          GTP-dependent polynucleotide kinase activity
          farnesol kinase activity
          CTP:2-trans,-6-trans-farnesol kinase activity
          geraniol kinase activity
          geranylgeraniol kinase activity
          CTP:geranylgeraniol kinase activity
          prenol kinase activity
          1-phosphatidylinositol-5-kinase activity
          1-phosphatidylinositol-3-phosphate 4-kinase activity
          phosphatidylinositol-3,4-bisphosphate 5-kinase activity
          phosphatidylinositol bisphosphate kinase activity
          inositol-3,4,6-trisphosphate 1-kinase activity
          inositol 5-diphosphate pentakisphosphate 5-kinase activity
          inositol diphosphate tetrakisphosphate kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline