Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ift43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199843.1 | NP_001186772.1 | 6 | 361 | ||
NM_029601.3 | NP_083877.1 | 5 | 328 | |||
GenBank mRNA | AK006115.1 | - | 5 | 326 | ||
BC034668.1 | - | 6 | 299 | |||
BC038915.1 | - | 6 | 350 | 5148723 | ||
CF751010.1 | - | 6 | 324 |