Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TH SIRNA Assays ›
          Gene Symbol:
          TH
          Gene Name:
          tyrosine hydroxylase
          Gene Aliases:
          DYT14, DYT5b, TYH
          Chromosome Location:
          Chr.11: 2163929 - 2174081 on Build GRCh38
          UniGene ID:
          Hs.435609
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 8425

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TH
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000360.3 NP_000351.2 10 1110
          NM_199292.2 NP_954986.2 11 1203
          NM_199293.2 NP_954987.2 11 1191
          XM_011520335.2 XP_011518637.1 10 3398
          GenBank mRNA AY144494.1 - 5 249
          AY144495.1 - 6 385
          BC104967.1 - 10 1182 8143970
          BC143611.1 - 10 1180 9052124
          BC143614.1 - 10 1194 9052127
          BM718799.1 - 7 565
          DQ677336.1 - 8 818
          DQ677337.1 - 8 830
          M17589.1 - 11 1193
          X05290.1 - 10 1110
          Y00414.1 - 11 1185

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          synaptic transmission, dopaminergic
          response to amphetamine
          heart morphogenesis
          dopamine biosynthetic process from tyrosine
          fatty acid metabolic process
          sphingolipid metabolic process
          heart development
          visual perception
          sensory perception of sound
          learning
          memory
          mating behavior
          locomotory behavior
          regulation of heart contraction
          response to water deprivation
          response to light stimulus
          response to herbicide
          response to salt stress
          anatomical structure morphogenesis
          organ morphogenesis
          response to zinc ion
          multicellular organism aging
          response to activity
          aminergic neurotransmitter loading into synaptic vesicle
          glycoside metabolic process
          phthalate metabolic process
          cerebral cortex development
          response to nutrient levels
          response to estradiol
          response to lipopolysaccharide
          isoquinoline alkaloid metabolic process
          social behavior
          cellular response to drug
          response to isolation stress
          response to immobilization stress
          neurotransmitter biosynthetic process
          terpene metabolic process
          dopamine biosynthetic process
          epinephrine biosynthetic process
          norepinephrine biosynthetic process
          catecholamine biosynthetic process
          eye photoreceptor cell development
          circadian sleep/wake cycle
          eating behavior
          response to peptide hormone
          pigmentation
          response to ethanol
          response to ether
          response to pyrethroid
          embryonic camera-type eye morphogenesis
          response to corticosterone
          response to electrical stimulus
          phytoalexin metabolic process
          oxidation-reduction process
          cellular response to manganese ion
          cellular response to nicotine
          cellular response to glucose stimulus
          cellular response to growth factor stimulus
          tyrosine 3-monooxygenase activity
          protein binding
          ferrous iron binding
          ferric iron binding
          amino acid binding
          oxygen binding
          enzyme binding
          protein domain specific binding
          tetrahydrobiopterin binding
          dopamine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0