Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029789.1 | NP_084065.1 | 7, 8 | 722 | ||
GenBank mRNA | AK002626.1 | - | 7, 8 | 689 | ||
AK050161.1 | - | 7, 8 | 703 | |||
AK146106.1 | - | 7, 8 | 701 | |||
AK159325.1 | - | 7, 8 | 701 | |||
AK206786.1 | - | 3, 4 | 199 | |||
AK216148.1 | - | 3, 4 | 176 | |||
AY029532.1 | - | 7, 8 | 722 | |||
BC006847.1 | - | 7, 8 | 694 | 3598473 |