Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prr15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030024.2 | NP_084300.1 | 2 | 720 | ||
GenBank mRNA | AJ132433.2 | - | 2 | 720 | ||
AK021401.1 | - | 2 | 828 | |||
AK136438.1 | - | 2 | 900 | |||
BC055838.1 | - | 2 | 849 | |||
BC132416.1 | - | 1 | 396 | 40130693 | ||
BC138757.1 | - | 1 | 396 | 8861779 |