Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030735.1 | NP_109660.1 | 2 | 503 | ||
GenBank mRNA | AF324873.1 | - | 2 | 503 | ||
BC139316.1 | - | 2 | 498 | 8862338 | ||
BC139317.1 | - | 2 | 498 | 8862339 | ||
BC145601.1 | - | 2 | 498 | 9054167 |