Search
Search

Tgfa| Gene Symbol: Tgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031199.3 | NP_112476.1 | 3 | 455 | ||
| GenBank mRNA | AK010785.1 | - | 3 | 483 | ||
| AK162680.1 | - | 3 | 220 | |||
| AK166321.1 | - | 3 | 388 | |||
| BC132185.1 | - | 3 | 253 | 40130462 | ||
| BC132211.1 | - | 3 | 253 | 40130488 | ||
| M92420.1 | - | 4 | 205 | |||
| U65016.1 | - | 3 | 290 | |||